Assay Details
Target Gene Details
Entrez Gene ID: | 11566 |
Gene Name: | adenylosuccinate synthetase, non muscle |
Gene Aliases: |
AI314886, AS, Adss2 |
Location: |
Chr.1:177763178-177796694 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adss | NM_007422.3 | 9 | 986 | NP_031448.2 |
| AK004877.1 | 9 | 939 | BAB23635.1 | |
| AK010263.1 | 9 | 961 | BAB26805.1 | |
| AK028060.1 | 9 | 986 | BAC25730.1 | |
| AK148420.1 | 9 | 968 | BAE28543.1 | |
| AK170279.1 | 9 | 972 | BAE41682.1 | |
| BC139404.1 | 9 | 898 | ||
| BC139417.1 | 9 | 898 | ||
| L24554.1 | 9 | 953 | AAA19727.1 |
More Information
Set Membership: |
|

Genomic Map