Assay Details
Target Gene Details
Entrez Gene ID: | 11650 |
Gene Name: | alkaline phosphatase, placental-like 2 |
Gene Aliases: |
Akp5, C77216, D1Ertd816e, EAP |
Location: |
Chr.1:87086690-87096250 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alppl2 | NM_007433.3 | NP_031459.3 | ||
| AK145571.1 | BAE26520.1 | |||
| AY054302.1 | AAL17657.1 | |||
| BC052874.1 | AAH52874.1 |
More Information
Set Membership: |
|

Genomic Map