Assay Details
Target Gene Details
Entrez Gene ID: | 252903 |
Gene Name: | adaptor-related protein complex AP-1, sigma 3 |
Gene Aliases: |
1190009B22, Jr2, [s]3A |
Location: |
Chr.1:79606876-79672020 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1s3 | NM_183027.2 | NP_898848.1 | ||
| AK028013.1 | ||||
| BC098237.1 | AAH98237.1 | |||
| BF715142.1 |
More Information
Set Membership: |
|

Genomic Map