Assay Details
Target Gene Details
Entrez Gene ID: | 52477 |
Gene Name: | angel homolog 2 (Drosophila) |
Gene Aliases: |
2610307I21Rik, 5730410O10Rik, AI845692, D1Ertd396e, D1Ertd654e |
Location: |
Chr.1:190925108-190946962 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angel2 | NM_001199020.1 | NP_001185949.1 | ||
| NM_021421.4 | NP_067396.3 | |||
| NR_037578.1 | ||||
| NR_037579.1 | ||||
| AB041602.1 | BAA95085.1 | |||
| AK077039.1 | ||||
| AK088834.1 | BAC40602.1 | |||
| AK135709.1 | BAE22620.1 | |||
| AK158731.1 | BAE34634.1 | |||
| AK166619.1 | BAE38898.1 | |||
| AK169653.1 | BAE41277.1 | |||
| AK178186.1 | ||||
| AK198112.1 | ||||
| AK207772.1 | ||||
| AK207828.1 | ||||
| AK209212.1 | ||||
| AK216227.1 | ||||
| BC024907.1 | AAH24907.1 |
More Information
Set Membership: |
|

Genomic Map