Assay Details
Target Gene Details
Entrez Gene ID: | 319695 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
4932422E22Rik |
Location: |
Chr.1:72642980-72700579 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankar | NM_176980.4 | 15 | 3146 | NP_795954.2 |
| AK030065.1 | 15 | 3147 | BAC26765.1 | |
| AK077036.1 | 14 | 2535 | ||
| BC132415.1 | 15 | 3121 | ||
| BC145253.1 | 15 | 3118 |

Genomic Map