Assay Details
Target Gene Details
Entrez Gene ID: | 319695 |
Gene Name: | ankyrin and armadillo repeat containing |
Gene Aliases: |
4932422E22Rik |
Location: |
Chr.1:72642980-72700579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankar | NM_176980.4 | NP_795954.2 | ||
| AK030065.1 | BAC26765.1 | |||
| AK077036.1 | ||||
| BC132415.1 | ||||
| BC145253.1 |

Genomic Map