Assay Details
Target Gene Details
Entrez Gene ID: | 65247 |
Gene Name: | ankyrin repeat and SOCS box-containing 1 |
Gene Aliases: |
1700029O08Rik, 1700054C17Rik, AI426616, AI851378, mKIAA1146 |
Location: |
Chr.1:91539459-91559590 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb1 | NM_001039126.2 | NP_001034215.1 | ||
| NM_023046.4 | NP_075533.3 | |||
| AF155352.1 | AAD38808.1 | |||
| AK006515.1 | BAB24628.1 | |||
| AK122456.1 | BAC65738.1 | |||
| AK153774.1 | BAE32180.1 | |||
| BC034869.1 | ||||
| BU938650.1 | ||||
| BY231875.1 | ||||
| KC789642.1 | ||||
| KC789643.1 |

Genomic Map