Assay Details
Target Gene Details
Entrez Gene ID: | 241158 |
Gene Name: | ankyrin repeat and MYND domain containing 1 |
Gene Aliases: |
4930483I10Rik, ZMYND13 |
Location: |
Chr.1:92859844-92902986 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankmy1 | NM_172850.3 | NP_766438.2 | ||
| AK029700.1 | BAC26572.1 | |||
| BC120887.1 | ||||
| BC120888.1 |

Genomic Map