Assay Details
Target Gene Details
Entrez Gene ID: | 404545 |
Gene Name: | anoctamin 7 |
Gene Aliases: |
Ngep, Ngep-L, Pcanap5, Tmem16g |
Location: |
Chr.1:93373796-93404304 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ano7 | NM_001271884.1 | NP_001258813.1 | ||
| NM_207031.2 | NP_996914.1 | |||
| BC116706.1 |

Genomic Map