Assay Details
Target Gene Details
Entrez Gene ID: | 108147 |
Gene Name: | 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase |
Gene Aliases: |
2610509C24Rik, AA536954, AW212393 |
Location: |
Chr.1:71557156-71579403 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atic | NM_026195.3 | NP_080471.2 | ||
| AK010449.1 | BAB26949.3 | |||
| AK010611.2 | BAB27060.1 | |||
| AK012074.1 | BAB28011.1 | |||
| AK028930.1 | 1 | 456 | ||
| AK046353.1 | BAC32688.1 | |||
| AK088701.1 | ||||
| AK139235.1 | BAE23927.1 | |||
| AK157739.1 | ||||
| BC039925.1 | AAH39925.2 | |||
| CJ178786.1 | ||||
| EF437353.1 |

Genomic Map