Assay Details
Target Gene Details
Entrez Gene ID: | 226562 |
Gene Name: | proline-rich coiled-coil 2C |
Gene Aliases: |
1810043M20Rik, 9630039I18Rik, A630006J20, Bat2d, Bat2d1, Bat2l2, E130112L15Rik, Prrc3, mKIAA1096 |
Location: |
Chr.1:162670727-162740564 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrc2c | NM_001081290.1 | NP_001074759.1 | ||
| AK041391.1 | ||||
| AK122441.1 | BAC65723.1 | |||
| BC050871.1 | AAH50871.1 |
More Information
Set Membership: |
|

Genomic Map