Assay Details
Target Gene Details
Entrez Gene ID: | 381306 |
Gene Name: | cDNA sequence BC055324 |
Gene Aliases: |
- |
Location: |
Chr.1:163945840-163994900 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC055324 | NM_201364.1 | 4 | 358 | NP_958752.1 |
| AK031594.1 | 4 | 353 | ||
| AK141661.1 | 4 | 362 | BAE24790.1 | |
| BC052855.1 | 4 | 352 | AAH52855.1 | |
| BC055324.1 | 4 | 358 | AAH55324.1 |

Genomic Map