Assay Details
Target Gene Details
Entrez Gene ID: | 69668 |
Gene Name: | coiled-coil domain containing 115 |
Gene Aliases: |
2310061I09Rik, Ccp1 |
Location: |
Chr.1:34436670-34439672 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc115 | NM_027159.2 | NP_081435.1 | ||
| AK010002.1 | BAB26634.1 | |||
| AK154179.1 | ||||
| AK162237.1 | BAE36808.1 | |||
| BC019430.1 | AAH19430.1 |

Genomic Map