Assay Details
Target Gene Details
Entrez Gene ID: | 78016 |
Gene Name: | coiled-coil domain containing 150 |
Gene Aliases: |
4930511H11Rik |
Location: |
Chr.1:54250661-54368727 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc150 | NM_030025.2 | NP_084301.2 | ||
| AK029994.1 | BAC26722.1 |

Genomic Map