Assay Details
Target Gene Details
Entrez Gene ID: | 11447 |
Gene Name: | cholinergic receptor, nicotinic, delta polypeptide |
Gene Aliases: |
Achr-4, Acrd, L10076 |
Location: |
Chr.1:87190597-87200070 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrnd | NM_021600.3 | 2 | 218 | NP_067611.2 |
| AK081344.1 | 1 | 204 | BAC38202.1 | |
| BC052153.1 | 2 | 163 | ||
| BC141376.1 | 2 | 146 | ||
| BC141387.1 | 2 | 146 | ||
| K02582.1 | 2 | 129 | AAA37157.1 | |
| L10076.1 | 2 | 178 | AAA39803.1 |
More Information
Set Membership: |
|

Genomic Map