Assay Details
Target Gene Details
Entrez Gene ID: | 11449 |
Gene Name: | cholinergic receptor, nicotinic, gamma polypeptide |
Gene Aliases: |
Achr-3, Acrg |
Location: |
Chr.1:87205811-87212410 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrng | NM_009604.3 | NP_033734.3 | ||
| AK047974.1 | ||||
| JF520813.1 | ||||
| JN164666.1 | ||||
| M30514.1 | AAB63431.1 | |||
| X03818.1 | CAA27442.1 | |||
| X03819.1 | CAA27443.1 |
More Information
Set Membership: |
|

Genomic Map