Assay Details
Target Gene Details
Entrez Gene ID: | 11449 |
Gene Name: | cholinergic receptor, nicotinic, gamma polypeptide |
Gene Aliases: |
Achr-3, Acrg |
Location: |
Chr.1:87205811-87212410 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrng | NM_009604.3 | 5 | 445 | NP_033734.3 |
| AK047974.1 | 5 | 446 | ||
| JF520813.1 | ||||
| JN164666.1 | 4 | 504 | ||
| M30514.1 | 5 | 486 | AAB63431.1 | |
| S77465.1 | 1 | 89 | AAB33997.2 | |
| X03818.1 | 5 | 442 | CAA27442.1 | |
| X03819.1 | 4 | 377 | CAA27443.1 |
More Information
Set Membership: |
|

Genomic Map