Assay Details
Target Gene Details
Entrez Gene ID: | 12790 |
Gene Name: | cyclic nucleotide gated channel alpha 3 |
Gene Aliases: |
CNG3 |
Location: |
Chr.1:37218278-37263384 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnga3 | NM_001282010.1 | NP_001268939.1 | ||
| NM_009918.2 | NP_034048.1 | |||
| AJ243933.1 | CAB89685.1 | |||
| BC035272.1 | AAH35272.1 | |||
| BC049145.1 | AAH49145.1 |
More Information
Set Membership: |
|

Genomic Map