Assay Details
Target Gene Details
Entrez Gene ID: | 12825 |
Gene Name: | collagen, type III, alpha 1 |
Gene Aliases: |
AW550625, Col3a-1, MMS10-W, Ms10w, mKIAA4231 |
Location: |
Chr.1:45311538-45349706 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col3a1 | NM_009930.2 | 2 | 397 | NP_034060.2 |
| AK013329.1 | 2 | 305 | BAB28793.1 | |
| AK029212.1 | 2 | 301 | ||
| AK031544.1 | 2 | 270 | ||
| AK041115.1 | 2 | 305 | BAC30826.1 | |
| AK048546.1 | 2 | 305 | BAC33370.1 | |
| AK147542.1 | 2 | 303 | BAE27985.1 | |
| AK157762.1 | 2 | 305 | BAE34185.1 | |
| AK159763.1 | 2 | 305 | ||
| AK159920.1 | 2 | 305 | ||
| AK159985.1 | 2 | 305 | ||
| AK160048.1 | 2 | 305 | ||
| AK160107.1 | 2 | 304 | BAE35633.1 | |
| AK160818.1 | 2 | 305 | ||
| AK220558.1 | 2 | 320 | BAD90325.1 | |
| BC043089.1 | 2 | 272 | AAH43089.1 | |
| BC052398.1 | 2 | 289 | AAH52398.1 | |
| BC058724.1 | 2 | 306 | AAH58724.1 | |
| BY139268.1 | ||||
| M18933.1 | 2 | 206 | AAA37338.1 |
More Information
Set Membership: |
|

Genomic Map