Assay Details
Target Gene Details
Entrez Gene ID: | 12825 |
Gene Name: | collagen, type III, alpha 1 |
Gene Aliases: |
AW550625, Col3a-1, MMS10-W, Ms10w, mKIAA4231 |
Location: |
Chr.1:45311538-45349706 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col3a1 | NM_009930.2 | 20 | 1612 | NP_034060.2 |
| AK029212.1 | 20 | 1515 | ||
| AK041115.1 | 20 | 1520 | BAC30826.1 | |
| AK048546.1 | 20 | 1520 | BAC33370.1 | |
| AK147542.1 | 20 | 1518 | BAE27985.1 | |
| AK157762.1 | 20 | 1520 | BAE34185.1 | |
| AK159763.1 | 20 | 1520 | ||
| AK159920.1 | 20 | 1520 | ||
| AK159985.1 | 20 | 1520 | ||
| AK160048.1 | 20 | 1520 | ||
| AK160107.1 | 20 | 1519 | BAE35633.1 | |
| AK160818.1 | 20 | 1520 | ||
| AK220558.1 | 20 | 1535 | BAD90325.1 | |
| BC028248.1 | 11 | 676 | AAH28248.1 | |
| BC043089.1 | 20 | 1487 | AAH43089.1 | |
| BC052398.1 | 20 | 1504 | AAH52398.1 | |
| BC058724.1 | 20 | 1521 | AAH58724.1 | |
| M18933.1 | 20 | 1421 | AAA37338.1 |
More Information
Set Membership: |
|

Genomic Map