Assay Details
Target Gene Details
Entrez Gene ID: | 12627 |
Gene Name: | cripto, FRL-1, cryptic family 1 |
Gene Aliases: |
AV265756, b2b970Clo, cryptic |
Location: |
Chr.1:34535648-34544311 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfc1 | NM_007685.2 | NP_031711.1 | ||
| BC100705.1 | AAI00706.1 | |||
| BC100706.1 | AAI00707.1 | |||
| BC100708.1 | AAI00709.1 | |||
| BC100711.1 | AAI00712.1 | |||
| U57720.1 | AAC53042.1 |
Target Gene Details
Entrez Gene ID: | 21756 |
Gene Name: | protease, serine 40 |
Gene Aliases: |
Tesp2 |
Location: |
Chr.1:34536515-34560993 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss40 |
More Information
Set Membership: |
|

Genomic Map