Assay Details
Target Gene Details
Entrez Gene ID: | 50702 |
Gene Name: | complement factor H-related 1 |
Gene Aliases: |
AI194696, CFHRB, Cfhl1 |
Location: |
Chr.1:139547064-139560222 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfhr1 | NM_015780.2 | 2 | 272 | NP_056595.1 |
| BC111870.1 | 2 | 255 | AAI11871.1 | |
| M29008.1 | 2 | 272 | AAA37414.1 |

Genomic Map