Assay Details
Target Gene Details
Entrez Gene ID: | 13244 |
Gene Name: | degenerative spermatocyte homolog 1 (Drosophila) |
Gene Aliases: |
AA536663, Degs, Des1, Mdes |
Location: |
Chr.1:182275770-182282759 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Degs1 | NM_007853.4 | NP_031879.1 | ||
| AF466376.1 | AAM12532.1 | |||
| AK002617.1 | BAB22233.1 | |||
| AK077257.1 | BAC36713.1 | |||
| AK149811.1 | BAE29098.1 | |||
| AK150908.1 | ||||
| AK163404.1 | BAE37337.1 | |||
| AK165352.1 | ||||
| BC003751.1 | AAH03751.1 | |||
| Y08460.1 | CAA69714.1 |
More Information
Set Membership: |
|

Genomic Map