Assay Details
Target Gene Details
Entrez Gene ID: | 170788 |
Gene Name: | crumbs family member 1, photoreceptor morphogenesis associated |
Gene Aliases: |
7530426H14Rik, A930008G09Rik |
Location: |
Chr.1:139197546-139379118 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crb1 | NM_133239.2 | 10 | 3974 | NP_573502.2 |
| AF406641.1 | 10 | 3939 | AAL65131.1 | |
| AF492496.1 | 9 | 3815 | AAQ06606.1 | |
| AK033059.1 | 2 | 87 | BAC28140.1 | |
| AK164413.1 | 2 | 938 | ||
| BC031378.1 | 3 | 1034 | AAH31378.1 | |
| BC053331.1 | 5 | 2217 | AAH53331.1 |
More Information
Set Membership: |
|

Genomic Map