Assay Details
Target Gene Details
Entrez Gene ID: | 170788 |
Gene Name: | crumbs family member 1, photoreceptor morphogenesis associated |
Gene Aliases: |
7530426H14Rik, A930008G09Rik |
Location: |
Chr.1:139197546-139379118 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crb1 | NM_133239.2 | NP_573502.2 | ||
| AF406641.1 | AAL65131.1 | |||
| AF492496.1 | AAQ06606.1 | |||
| BC031378.1 | AAH31378.1 | |||
| BC053331.1 | AAH53331.1 |
More Information
Set Membership: |
|

Genomic Map