Assay Details
Target Gene Details
Entrez Gene ID: | 12912 |
Gene Name: | cAMP responsive element binding protein 1 |
Gene Aliases: |
2310001E10Rik, 3526402H21Rik, AV083133, Creb, Creb-1 |
Location: |
Chr.1:64532804-64604548 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creb1 | NM_001037726.1 | NP_001032815.1 | ||
| NM_009952.2 | NP_034082.1 | |||
| NM_133828.2 | NP_598589.2 | |||
| AF448507.1 | AAL47130.1 | |||
| AK042595.1 | ||||
| AK044992.1 | BAC32173.1 | |||
| AK044993.1 | BAC32174.1 | |||
| AK079223.1 | ||||
| BC021649.1 | AAH21649.1 | |||
| BY315899.1 | ||||
| BY782293.1 | ||||
| M95106.1 | AAA37456.1 | |||
| U46027.1 | AAB64015.1 | |||
| X92497.1 | CAA63242.1 |

Genomic Map