Assay Details
Target Gene Details
Entrez Gene ID: | 12477 |
Gene Name: | cytotoxic T-lymphocyte-associated protein 4 |
Gene Aliases: |
Cd152, Ctla-4, Ly-56 |
Location: |
Chr.1:60909025-60915832 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctla4 | NM_001281976.1 | NP_001268905.1 | ||
| NM_009843.4 | NP_033973.2 | |||
| AB097214.1 | BAC41333.1 | |||
| AK138891.1 | ||||
| BC042741.1 | AAH42741.1 | |||
| BC052683.1 | AAH52683.1 | |||
| U90270.1 | AAD00696.1 | |||
| X05719.1 | CAA29191.1 |
More Information
Set Membership: |
|

Genomic Map