Assay Details
Target Gene Details
Entrez Gene ID: | 13800 |
Gene Name: | enabled homolog (Drosophila) |
Gene Aliases: |
Mena, NDPP-1, Ndpp1, WBP8 |
Location: |
Chr.1:181896384-182019980 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enah | NM_001083120.2 | NP_001076589.1 | ||
| NM_001083121.2 | NP_001076590.1 | |||
| NM_008680.3 | NP_032706.2 | |||
| NM_010135.3 | NP_034265.2 | |||
| BC062927.1 | AAH62927.1 | |||
| U72520.1 | AAC52863.1 | |||
| U72521.1 | AAC52864.1 | |||
| U72522.1 | AAC52865.1 | |||
| U72523.1 | AAC52866.1 |
More Information
Set Membership: |
|

Genomic Map