Assay Details
Target Gene Details
Entrez Gene ID: | 14200 |
Gene Name: | four and a half LIM domains 2 |
Gene Aliases: |
FHL-2, SLIM-3, SLIM3 |
Location: |
Chr.1:43123071-43198299 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhl2 | NM_001289533.1 | NP_001276462.1 | ||
| NM_010212.4 | NP_034342.1 | |||
| AF055889.1 | AAC12770.1 | |||
| AF114381.1 | AAD53230.1 | |||
| AF153340.1 | AAD34170.1 | |||
| AK052936.1 | BAC35211.1 | |||
| AK133708.1 | ||||
| AK147024.1 | BAE27617.1 | |||
| BC021468.1 | AAH21468.1 | |||
| U77040.2 | AAB19211.2 |
More Information
Set Membership: |
|

Genomic Map