Assay Details
Target Gene Details
Entrez Gene ID: | 26909 |
Gene Name: | exonuclease 1 |
Gene Aliases: |
5730442G03Rik, Msa |
Location: |
Chr.1:175880761-175911396 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exo1 | NM_012012.4 | 3 | 229 | NP_036142.2 |
| AJ238213.1 | 1 | 84 | CAB51863.1 | |
| AK028728.1 | 3 | 210 | BAC26086.1 | |
| AK166425.1 | 3 | 205 | BAE38768.1 | |
| BC006671.1 | 3 | 193 | AAH06671.1 |
More Information
Set Membership: |
|

Genomic Map