Assay Details
Target Gene Details
Entrez Gene ID: | 63913 |
Gene Name: | family with sequence similarity 129, member A |
Gene Aliases: |
AI256368, AU019833, Niban |
Location: |
Chr.1:151571373-151721939 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam129a | NM_022018.3 | 5 | 670 | NP_071301.2 |
| AB049355.1 | 5 | 510 | BAB17052.1 | |
| AK136597.1 | 5 | 580 | BAE23066.1 | |
| AK170193.1 | 5 | 593 | BAE41626.1 | |
| BC021332.1 | 5 | 558 | ||
| BC058234.1 | 3 | 270 | ||
| BC137843.1 | 1 | 47 |

Genomic Map