Assay Details
Target Gene Details
Entrez Gene ID: | 63913 |
Gene Name: | family with sequence similarity 129, member A |
Gene Aliases: |
AI256368, AU019833, Niban |
Location: |
Chr.1:151571373-151721939 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam129a | NM_022018.3 | 7 | 922 | NP_071301.2 |
| AB049355.1 | 7 | 762 | BAB17052.1 | |
| AK029847.1 | 3 | 234 | BAC26641.1 | |
| AK136597.1 | 7 | 832 | BAE23066.1 | |
| AK170193.1 | 7 | 845 | BAE41626.1 | |
| BC021332.1 | 7 | 810 | ||
| BC058234.1 | 5 | 522 | ||
| BC137843.1 | 3 | 299 |

Genomic Map