Assay Details
Target Gene Details
Entrez Gene ID: | 227298 |
Gene Name: | family with sequence similarity 134, member A |
Gene Aliases: |
- |
Location: |
Chr.1:75142786-75147909 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam134a | NM_170755.2 | NP_739561.2 | ||
| AK034667.1 | BAC28789.1 | |||
| AK152268.1 | ||||
| AK158940.1 | ||||
| AK160400.1 | BAE35769.1 | |||
| AK165957.1 | BAE38485.1 | |||
| AK172093.1 | BAE42822.1 | |||
| AK189209.1 | ||||
| AK191491.1 | ||||
| AK210282.1 | ||||
| AK210397.1 | ||||
| AK210767.1 | ||||
| AK218481.1 | ||||
| BC026519.1 | AAH26519.1 | |||
| BC038286.1 | AAH38286.1 | |||
| BC070406.1 | AAH70406.1 |

Genomic Map