Assay Details
Target Gene Details
Entrez Gene ID: | 14633 |
Gene Name: | GLI-Kruppel family member GLI2 |
Gene Aliases: |
AW546128 |
Location: |
Chr.1:118834061-119053619 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gli2 | NM_001081125.1 | 3 | 536 | NP_001074594.1 |
| AM261668.1 | ||||
| AM261669.1 | ||||
| AM261670.1 | 3 | 256 | ||
| AM262741.1 | 3 | 256 | ||
| AM262742.1 | 3 | 256 | ||
| AM262743.1 | ||||
| AM262744.1 | 4 | 367 | ||
| BC082604.1 | 3 | 477 | ||
| BC085190.1 | 3 | 452 | ||
| X99104.1 | 3 | 296 |
More Information
Set Membership: |
|

Genomic Map