Assay Details
Target Gene Details
Entrez Gene ID: | 68187 |
Gene Name: | family with sequence similarity 135, member A |
Gene Aliases: |
4921533L14Rik, AI256732, AI835491 |
Location: |
Chr.1:24010758-24100460 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam135a | NM_026604.4 | NP_080880.4 | ||
| AK053786.1 | ||||
| AK081262.1 | ||||
| AK149303.1 | BAE28800.1 | |||
| AK165306.1 | ||||
| BC070446.1 | AAH70446.1 |

Genomic Map