Assay Details
Target Gene Details
Entrez Gene ID: | 381337 |
Gene Name: | family with sequence similarity 178, member B |
Gene Aliases: |
1700024G10Rik, Gm990 |
Location: |
Chr.1:36562696-36683190 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam178b | NM_001126046.1 | 14 | 1923 | NP_001119518.1 |
| NM_027957.1 | 5 | 546 | NP_082233.1 | |
| AK006300.1 | 5 | 547 | BAB24515.1 | |
| BC114576.1 | 5 | 465 | AAI14577.1 | |
| BU605266.1 | 2 | 172 | ||
| BY445320.1 | 1 | 60 |

Genomic Map