Assay Details
Target Gene Details
Entrez Gene ID: | 215015 |
Gene Name: | family with sequence similarity 20, member B |
Gene Aliases: |
C530043G21Rik, mKIAA0475 |
Location: |
Chr.1:156678545-156719027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam20b | NM_145413.4 | NP_663388.1 | ||
| AK049705.1 | BAC33883.1 | |||
| AK129154.1 | BAC97964.1 | |||
| AK132149.1 | BAE20998.1 | |||
| AK182974.1 | ||||
| AK190376.1 | ||||
| BC019381.1 | AAH19381.1 | |||
| BC023737.1 | AAH23737.1 | |||
| BC024412.1 | AAH24412.1 | |||
| BC031473.1 | AAH31473.1 |

Genomic Map