Assay Details
Target Gene Details
Entrez Gene ID: | 227399 |
Gene Name: | diphosphoinositol pentakisphosphate kinase 2 |
Gene Aliases: |
AW555814, D330021B20, Hisppd1, Vip2, mKIAA0433 |
Location: |
Chr.1:97706043-97770250 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppip5k2 | NM_173760.5 | NP_776121.4 | ||
| AK087712.1 | ||||
| AK129140.1 | BAC97950.1 | |||
| AK138622.1 | BAE23724.1 | |||
| AK149804.1 | ||||
| AK162179.1 | ||||
| BC046411.1 | AAH46411.1 | |||
| BC053396.1 | AAH53396.1 |
More Information
Set Membership: |
|

Genomic Map