Assay Details
Target Gene Details
Entrez Gene ID: | 14859 |
Gene Name: | glutathione S-transferase, alpha 3 |
Gene Aliases: |
Gst2-3 |
Location: |
Chr.1:21240585-21265575 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsta3 | NM_001077353.2 | NP_001070821.1 | ||
| NM_001288617.1 | NP_001275546.1 | |||
| NM_010356.4 | NP_034486.2 | |||
| AI047326.1 | ||||
| AI255699.1 | ||||
| AK002478.1 | BAB22131.1 | |||
| AK014076.1 | BAB29143.1 | |||
| AK149543.1 | BAE28948.1 | |||
| BC147272.1 | ||||
| BC147273.1 | ||||
| GQ227791.1 | ||||
| M73483.1 | AAA37751.1 | |||
| X65021.1 | CAA46155.1 |
More Information
Set Membership: |
|

Genomic Map