Assay Details
Target Gene Details
Entrez Gene ID: | 381314 |
Gene Name: | isoleucine-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
2010002H18Rik, C79125 |
Location: |
Chr.1:185286662-185329419 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iars2 | NM_198653.2 | 11 | 1511 | NP_941055.1 |
| AK036182.1 | 11 | 1479 | BAC29337.1 | |
| AK043590.1 | 11 | 1513 | ||
| AK043729.1 | 11 | 1489 | BAC31631.1 | |
| AK048653.1 | 11 | 1512 | BAC33410.1 | |
| AK050358.1 | 11 | 1494 | BAC34208.1 | |
| AK051778.1 | 11 | 1472 | ||
| AK078183.1 | 11 | 1513 | ||
| AK085714.1 | 11 | 1508 | ||
| AK088965.1 | 11 | 1472 | ||
| AK166832.1 | 11 | 1494 | BAE39055.1 | |
| BC052403.1 | 11 | 1488 | AAH52403.1 |
More Information
Set Membership: |
|

Genomic Map