Assay Details
Target Gene Details
Entrez Gene ID: | 74918 |
Gene Name: | IQ motif containing with AAA domain |
Gene Aliases: |
4930465P12Rik, 4930585L22Rik, BB288505, Iqca1 |
Location: |
Chr.1:90042132-90153401 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqca | NM_029122.2 | 15 | 2044 | NP_083398.2 |
| AK139726.1 | 8 | 899 | BAE24115.1 |

Genomic Map