Assay Details
Target Gene Details
Entrez Gene ID: | 16178 |
Gene Name: | interleukin 1 receptor, type II |
Gene Aliases: |
CD121b, Il1r-2 |
Location: |
Chr.1:40081750-40125230 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1r2 | NM_010555.4 | 8 | 1124 | NP_034685.1 |
| AK132264.1 | 8 | 1151 | BAE21067.1 | |
| BC032962.1 | 7 | 841 | AAH32962.1 | |
| CT010182.1 | 7 | 1032 | CAJ18390.1 | |
| X59769.1 | 8 | 1124 | CAA42440.1 |
More Information
Set Membership: |
|

Genomic Map