Assay Details
Target Gene Details
Entrez Gene ID: | 117198 |
Gene Name: | influenza virus NS1A binding protein |
Gene Aliases: |
1190004M08Rik, 1700126I16Rik, AA960440, HSPC068, ND1, NS-1, NS1-BP, Nd1-L, Nd1-S, mKIAA0850 |
Location: |
Chr.1:151344308-151364445 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ivns1abp | NM_001039512.1 | NP_001034601.1 | ||
| NM_054102.2 | NP_473443.2 | |||
| AB055737.1 | BAB69058.1 | |||
| AK129229.1 | BAC98039.1 | |||
| AK144619.1 | ||||
| AK144835.1 | BAE26089.1 | |||
| AK145071.1 | BAE26221.1 | |||
| AK145845.1 | ||||
| AK146001.1 | BAE26822.1 | |||
| AK146329.1 | BAE27084.1 | |||
| AK146494.1 | BAE27212.1 | |||
| AK146645.1 | BAE27326.1 | |||
| AK159254.1 | BAE34935.1 | |||
| AK197779.1 | ||||
| AK204006.1 | ||||
| BC004092.1 | AAH04092.1 | |||
| DQ914522.1 | ||||
| DQ914523.1 |
More Information
Set Membership: |
|

Genomic Map