Assay Details
Target Gene Details
Entrez Gene ID: | 107527 |
Gene Name: | interleukin 1 receptor-like 2 |
Gene Aliases: |
AI481289, AW551444, IL-1Rrp2 |
Location: |
Chr.1:40324441-40367585 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rl2 | NM_133193.3 | NP_573456.1 | ||
| AF284433.1 | AAG21367.1 | |||
| AK085780.1 | ||||
| BC117797.2 |
More Information
Set Membership: |
|

Genomic Map