Assay Details
Target Gene Details
Entrez Gene ID: | 16526 |
Gene Name: | potassium channel, subfamily K, member 2 |
Gene Aliases: |
A430027H14Rik, AI848635, TREK-1 |
Location: |
Chr.1:189207930-189402705 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk2 | NM_001159850.1 | NP_001153322.1 | ||
| NM_001281847.1 | NP_001268776.1 | |||
| NM_001281848.1 | NP_001268777.1 | |||
| NM_010607.3 | NP_034737.2 | |||
| AK138427.1 | BAE23658.1 | |||
| AK140716.1 | BAE24455.1 | |||
| AK164197.1 | ||||
| AY736359.1 | AAV48996.1 | |||
| BC062094.1 | AAH62094.1 | |||
| CN528767.1 | ||||
| U73488.2 | AAC53005.2 |
More Information
Set Membership: |
|

Genomic Map