Assay Details
Target Gene Details
Entrez Gene ID: | 269181 |
Gene Name: | mannoside acetylglucosaminyltransferase 4, isoenzyme A |
Gene Aliases: |
9530018I07Rik, GnT-IVa, glcNAc-T-IVa |
Location: |
Chr.1:37439340-37541047 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat4a | NM_001290801.1 | 2 | 446 | NP_001277730.1 |
| NM_173870.3 | 3 | 498 | NP_776295.1 | |
| AB053217.1 | 2 | 174 | BAC55018.1 | |
| AK155724.1 | 3 | 499 | ||
| AK163327.1 | 2 | 278 | BAE37301.1 |
More Information
Set Membership: |
|

Genomic Map