Assay Details
Target Gene Details
Entrez Gene ID: | 280287 |
Gene Name: | KiSS-1 metastasis-suppressor |
Gene Aliases: |
kisspeptin, metastatin |
Location: |
Chr.1:133321887-133329722 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kiss1 | NM_178260.3 | NP_839991.2 | ||
| AA023297.1 | ||||
| AB666166.1 | 2 | 143 | ||
| AF472576.1 | AAP13356.1 | |||
| BC117046.1 | ||||
| BC119348.1 |

Genomic Map