Assay Details
Target Gene Details
Entrez Gene ID: | 26921 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 4 |
Gene Aliases: |
9430080K19Rik, AU043147, AU045934, AW046177, HGK, Nik |
Location: |
Chr.1:39900912-40026310 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map4k4 | NM_001252200.1 | NP_001239129.1 | ||
| NM_001252201.1 | NP_001239130.1 | |||
| NM_001252202.1 | NP_001239131.1 | |||
| NM_008696.2 | NP_032722.2 | |||
| AK129191.1 | BAC98001.1 | |||
| BC141109.1 | ||||
| BC145390.1 | ||||
| BC145391.1 | ||||
| CF531764.1 | ||||
| U88984.1 | AAC53165.1 |
More Information
Set Membership: |
|

Genomic Map