Assay Details
Target Gene Details
Entrez Gene ID: | 213006 |
Gene Name: | major facilitator superfamily domain containing 4 |
Gene Aliases: |
A230072B04, A930031D07Rik, AI850289 |
Location: |
Chr.1:132022806-132068062 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd4 | NM_001114662.1 | NP_001108134.1 | ||
| NM_172510.4 | NP_766098.2 | |||
| AK038882.1 | BAC30158.1 | |||
| AK044667.1 | ||||
| AK048692.1 | BAC33423.1 | |||
| AK085484.1 | BAC39455.1 | |||
| AK162495.1 | BAE36946.1 | |||
| BC058790.1 | AAH58790.1 | |||
| BC080198.1 | AAH80198.1 |
More Information
Set Membership: |
|

Genomic Map