Assay Details
Target Gene Details
Entrez Gene ID: | 17970 |
Gene Name: | neutrophil cytosolic factor 2 |
Gene Aliases: |
NOXA2, Ncf-2, p67phox |
Location: |
Chr.1:152800158-152837169 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncf2 | NM_010877.4 | NP_035007.1 | ||
| AB002664.1 | BAA25650.1 | |||
| AK036379.1 | BAC29404.1 | |||
| AK137152.1 | BAE23254.1 | |||
| AK153447.1 | BAE32002.1 | |||
| AK155324.1 | BAE33191.1 | |||
| AK170839.1 | BAE42065.1 | |||
| AK171059.1 | BAE42218.1 | |||
| AK171265.1 | BAE42353.1 | |||
| BC003730.1 | AAH03730.1 | |||
| DQ449939.1 | ABE02824.1 |
More Information
Set Membership: |
|

Genomic Map